Learn More
Invitrogen™ RFC2 Monoclonal Antibody (OTI3A7)
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA526417
Product Code. 15946014
  • £474.00 / 100µL
Description
RFC2 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
This gene encodes a member of the activator 1 small subunits family. The elongation of primed DNA templates by DNA polymerase delta and epsilon requires the action of the accessory proteins, proliferating cell nuclear antigen (PCNA) and replication factor C (RFC). Replication factor C, also called activator 1, is a protein complex consisting of five distinct subunits. This gene encodes the 40 kD subunit, which has been shown to be responsible for binding ATP and may help promote cell survival. Disruption of this gene is associated with Williams syndrome. Alternatively spliced transcript variants encoding distinct isoforms have been described. A pseudogene of this gene has been defined on chromosome 2.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RFC2
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
P35250
RFC2
Human recombinant protein fragment corresponding to amino acids 1-234 of RFC2 produced in E.coli.
100 μL
Primary
Human
Antibody
IgG1
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
OTI3A7
Unconjugated
RFC2
2610008M13Rik; 40kDa; A1 40 kDa subunit; Activator 1 40 kDa subunit; activator 1 subunit 2; Activator 1 subunit C2; AI326953; MGC3665; Recc2; replication factor C (activator 1) 2; replication factor C (activator 1) 2, 40kDa; replication factor C 40 kDa subunit; Replication factor C subunit 2; replication factor C, 40 kDa; RF-C 40 kDa subunit; RFC2; RFC40
Mouse
Affinity Chromatography
RUO
5982
-20°C, Avoid Freeze/Thaw Cycles