missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RFC2 Antibody (OTI4C11), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73858DL594
This item is not returnable.
View return policy
Description
RFC2 Monoclonal antibody specifically detects RFC2 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry
Specifications
| RFC2 | |
| Monoclonal | |
| DyLight 594 | |
| A1, A1 40 kDa subunit, Activator 1 40 kDa subunit, Activator 1 subunit 2, MGC3665, replication factor C (activator 1) 2, 40kDa, replication factor C 2, Replication factor C 40 kDa subunit, replication factor C subunit 2, RF-C 40 kDa subunit, RFC40replication factor C (activator 1) 2 (40kD) | |
| Recombinant protein fragment corresponding to amino acids 1-234 of human RFC2 (NP_002905) produced in E.coli. | |
| 0.1 mL | |
| Cell Cycle and Replication, DNA Repair | |
| 5982 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| OTI4C11 | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction