missing translation for 'onlineSavingsMsg'
Learn More

RFC2 Antibody, HRP, Novus Biologicals™

Product Code. 18414289 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18414289 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18414289 Supplier Novus Biologicals Supplier No. NB100231H

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

RFC2 Polyclonal antibody specifically detects RFC2 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen RFC2
Applications Western Blot
Classification Polyclonal
Conjugate HRP
Dilution Western Blot
Formulation PBS
Gene Alias A1, A1 40 kDa subunit, Activator 1 40 kDa subunit, Activator 1 subunit 2, MGC3665, replication factor C (activator 1) 2, 40kDa, replication factor C 2, Replication factor C 40 kDa subunit, replication factor C subunit 2, RF-C 40 kDa subunit, RFC40replication factor C (activator 1) 2 (40kD)
Host Species Goat
Immunogen The immunogen recognized by this antibody maps to a region between residues 1 and 50 of human Replication Factor C subunit 2 using the numbering given in entry NP_852136.1 (GeneID 5982).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 5982
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.