missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RFC2 Antibody, CoraFluor™ 1, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB100-231CL1
This item is not returnable.
View return policy
Description
RFC2 Polyclonal antibody specifically detects RFC2 in Human samples. It is validated for Western Blot
Specifications
| RFC2 | |
| Polyclonal | |
| PBS | |
| Goat | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| CoraFluor 1 | |
| A1, A1 40 kDa subunit, Activator 1 40 kDa subunit, Activator 1 subunit 2, MGC3665, replication factor C (activator 1) 2, 40kDa, replication factor C 2, Replication factor C 40 kDa subunit, replication factor C subunit 2, RF-C 40 kDa subunit, RFC40replication factor C (activator 1) 2 (40kD) | |
| The immunogen recognized by this antibody maps to a region between residues 1 and 50 of human Replication Factor C subunit 2 using the numbering given in entry NP_852136.1 (GeneID 5982). | |
| 0.1 mL | |
| Cell Cycle and Replication, DNA Repair | |
| 5982 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction