Learn More
Heathrow Scientific™ Rechargeable Battery
Product Code. 12924417 Shop All Thermo Scientific CO₂ Incubators Products
Click to view available options
:
Battery Type: Lithium
Product Code. 12924417
Brand: Heathrow Scientific™ HS300502
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Charges quickly for use up to 20 hours of continuous use. Heathrow Scientific™ Rechargeable Battery for RF3000™ Pipette Controller provides 100 to 240 volts.
  • For use with Heathrow Scientific™ RF3000 Pipette Controller.
  • Provides 100 to 240 volt electrical supplies worldwide.
  • Long-life li-ion battery, with quick charge–up to 20 hours continuous use.
  • Low battery indicator; can be recharged when in use
Specifications
Battery Type Lithium-ion
Model RF3000
Type Rechargeable Battery
For Use With (Equipment) RF3000™ Pipette Controller
Quantity 1 Pack
Voltage 100 to 240 V