missing translation for 'onlineSavingsMsg'
Learn More

Reynoutrin, MedChemExpress

Product Code. 16494907
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16494907 50 mg 50mg
16489208 10 mg 10mg
16431628 5 mg 5mg
16423916 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16494907 Supplier Medchem Express Supplier No. HYN1354

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Reynoutrin (Quercetin-3-D-xyloside) is a flavonoid from Psidium cattleianum, with antioxidant and radical-scavenging activity.

Reynoutrin (Quercetin-3-D-xyloside) is a flavonoid from Psidium cattleianum, with antioxidant and radical-scavenging activity.

Specifications

Chemical Name or Material Reynoutrin
CAS 549-32-6
Color Yellow
Purity Grade Notes Research
Molecular Formula C20H18O11
Quantity 50 mg
Synonym Quercetin-3-D-xyloside Reinutrin
Solubility Information DMSO : 150 mg/mL (345.34 mM; Need ultrasonic)
SMILES O=C1C(O[C@H]2[C@@H]([C@H]([C@@H](CO2)O)O)O)=C(C3=CC=C(O)C(O)=C3)OC4=CC(O)=CC(O)=C14
Molecular Weight (g/mol) 434.35
Formula Weight 434.35
Percent Purity 97.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.