Learn More
Invitrogen™ Rex1 Recombinant Rabbit Monoclonal Antibody (14H16L9)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ 701159
Description
Intact IgG appears on a non-reducing gel as ~150 kDa band and upon reduction generating a ~25 kDa light chain band and a ~50 kDa heavy chain. Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
Involved in the reprogramming of X-chromosome inactivation during the acquisition of pluripotency. Required for efficient elongation of TSIX, a non-coding RNA antisense to XIST. Binds DXPas34 enhancer within the TSIX promoter. Involved in ES cell self-renewal. [UniProt].
Specifications
| Rex1 | |
| Recombinant Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide | |
| Q96MM3 | |
| ZFP42 | |
| Recombinant protein corresponding to amino acids 1-140 of human REX1 transcription factor. | |
| 100 μg | |
| Primary | |
| Human, Canine | |
| Antibody | |
| IgG |
| Western Blot, Immunocytochemistry | |
| 14H16L9 | |
| Unconjugated | |
| ZFP42 | |
| 1700021P10Rik; 2610511M11Rik; AW050347; ELOABP1; EloA-BP1; elongin A binding protein 1; elongin A-binding protein 1; elongin-A-binding protein 1; hREX-1; KIAA1138; mREX-1; reduced expression protein 1; REX1; Rex-1; REX1 transcription factor; REX1, RNA exonuclease 1 homolog; REX1, RNA exonuclease 1 homolog (S. cerevisiae); REXO1; RNA exonuclease 1 homolog; Tceb3bp1; transcription elongation factor B polypeptide 3 binding protein 1; transcription elongation factor B polypeptide 3-binding protein 1; Zfp42; Zfp-42; ZFP42 zinc finger protein; Zfp971; Zinc finger protein 42; zinc finger protein 42 homolog; Zinc finger protein 754; zinc finger protein 971; ZNF754 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 100856057, 132625 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.