missing translation for 'onlineSavingsMsg'
Learn More

Rex-1/ZFP42 Antibody (5E11E7) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

£161.00 - £431.00

Specifications

Antigen Rex-1/ZFP42
Clone 5E11E7
Concentration 1 mg/mL
Dilution Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000, CyTOF-ready
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18690516
View Documents
Novus Biologicals
NBP2-37360-0.025ml
0.025 mL
£161.00
0.03mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18159797
View Documents
Novus Biologicals
NBP2-37360
0.1 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Rex-1/ZFP42 Monoclonal specifically detects Rex-1/ZFP42 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, CyTOF-ready.
Specifications

Specifications

Rex-1/ZFP42
1 mg/mL
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF
Unconjugated
RUO
PBS with 0.05% Sodium Azide
132625
Purified recombinant fragment of human ZFP42 (AA: 249-310) expressed in E. Coli.
Primary
34.8 kDa
5E11E7
Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000, CyTOF-ready
Monoclonal
Mouse
Human
FLJ20086, KIAA1584, MGC21637, MGC61687, SUHW4, Suppressor of hairy wing homolog 4, zinc finger protein 280D, Zinc finger protein 634, ZNF634suppressor of hairy wing homolog 4 (Drosophila)
ZFP42
IgG1
Affinity Purified
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.