missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Revdofilimab Humanized Recombinant Human Monoclonal Antibody
Human Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA554975
This item is not returnable.
View return policy
Description
Compatible with Direct ELISA.
Revdofilimab is a monoclonal antibody that targets and inhibits CD40, a co-stimulatory protein on antigen-presenting cells, thereby modulating immune responses.
Specifications
| Revdofilimab Humanized | |
| Recombinant Monoclonal | |
| Unconjugated | |
| C161864 | |
| Recombinant Human CD134/TNFRSF4/OX40. | |
| 100 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA | |
| 3 mg/mL | |
| PBS with no preservative | |
| Human | |
| Protein A/G | |
| RUO | |
| Human | |
| Antibody | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction