missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ REV1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16637354
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16637354 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16637354 Supplier Invitrogen™ Supplier No. PA5100870

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total REV1.

This gene encodes a protein with similarity to the S. cerevisiae mutagenesis protein Rev1. The Rev1 proteins contain a BRCT domain, which is important in protein-protein interactions. A suggested role for the human Rev1-like protein is as a scaffold that recruits DNA polymerases involved in translesion synthesis (TLS) of damaged DNA. Two alternatively spliced transcript variants that encode different proteins have been found.
TRUSTED_SUSTAINABILITY

Specifications

Antigen REV1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Rev1
Gene Accession No. Q920Q2, Q9UBZ9
Gene Alias 1110027I23Rik; AIBP80; alpha integrin-binding protein 80; AU022044; DNA repair protein REV1; REV1; REV1 homolog; REV1 homolog (S. cerevisiae); REV1- like; REV1, DNA directed polymerase; REV1, polymerase (DNA directed); REV1L; rev1-like terminal deoxycytidyl transferase
Gene Symbols Rev1
Host Species Rabbit
Immunogen A synthesized peptide derived from human REV1(Accession Q9UBZ9), corresponding to amino acid residues Q1219-T1251.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51455, 56210
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.