missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rev-erb beta/NR1D2 Antibody (OTI5E10), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-73854CL1
This item is not returnable.
View return policy
Description
Rev-erb beta/NR1D2 Monoclonal antibody specifically detects Rev-erb beta/NR1D2 in Human,Mouse,Rat samples. It is validated for Western Blot
Specifications
| Rev-erb beta/NR1D2 | |
| Monoclonal | |
| CoraFluor 1 | |
| BD73, EAR-1r, HZF2, nuclear receptor subfamily 1 group D member 2, nuclear receptor subfamily 1, group D, member 2, Orphan nuclear hormone receptor BD73, rev-erba-alpha-related receptor, rev-erb-beta, RVR, V-erbA-related protein 1-related | |
| Human recombinant protein fragment corresponding to amino acids 1-294 of human NR1D2(NP_005117) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| OTI5E10 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 9975 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction