missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rev-erb beta/NR1D2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-50165
This item is not returnable.
View return policy
Description
Rev-erb beta/NR1D2 Polyclonal antibody specifically detects Rev-erb beta/NR1D2 in Human, Mouse, Rat, Bat, Bovine, Canine, Chicken, Equine, Primate, Monkey, Rabbit samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Rev-erb beta/NR1D2 | |
| Polyclonal | |
| Unconjugated | |
| PBS | |
| BD73, EAR-1r, HZF2, nuclear receptor subfamily 1 group D member 2, nuclear receptor subfamily 1, group D, member 2, Orphan nuclear hormone receptor BD73, rev-erba-alpha-related receptor, rev-erb-beta, RVR, V-erbA-related protein 1-related | |
| Synthetic 16 amino acid peptide from internal region of human Rev-erb beta/NR1D2. | |
| 0.05 mL | |
| Primary | |
| Human, Mouse, Rat, Bat, Bovine | |
| Purified |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| ELISA 1:100 - 1:2000, Immunohistochemistry, Immunohistochemistry-Paraffin 10 μg/mL | |
| Q14995 | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 9975 | |
| Keep as concentrated solution. Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction