missing translation for 'onlineSavingsMsg'
Learn More

Rev-erb A alpha/NR1D1 Antibody, Novus Biologicals™

Product Code. 18420801 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
25 μL
Unit Size:
0.10mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18420801 25 μL 25µL
18449440 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18420801 Supplier Novus Biologicals Supplier No. NBP18493125ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Rev-erb A alpha/NR1D1 Polyclonal antibody specifically detects Rev-erb A alpha/NR1D1 in Human, Amphibian samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Frozen)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rev-erb A alpha/NR1D1
Applications Immunocytochemistry, Immunohistochemistry (Frozen)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL, Immunohistochemistry-Frozen -Validated from a verified customer review.
Formulation PBS (pH 7.2) and 40% Glycerol
Gene Alias EAR-1, EAR1nuclear receptor subfamily 1 group D member 1, HREV, nuclear receptor subfamily 1, group D, member 1, Rev-erbA-alpha, Rev-ErbAalpha, THRA1, THRALrev-erbA-alpha, V-erbA-related protein 1
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids: TTLDSNNNTGGVITYIGSSGSSPSRTSPESLYSDNSNGSFQSLTQGCPTYFPPSPTGSLTQDPARSFGSIPPSLSDDGSPSSSSSSSSSSSSFYNGSPPGSLQVAMEDSSRVSPSKSTSNITKLNGMVLLCKVCGDVA
Purification Method Immunogen affinity purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9572
Target Species Human, Amphibian
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.