Learn More
3M™ Reusable Half Mask Respirator Kit A1P2 R
Product Code. 15759797
Change view
Click to view available options
Size: Large
Product Code.
15759797 Size: Large
1 options
Product Code. 15759797 Supplier 3M™ Supplier No. 7000061548
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
3M™ Reusable Half Mask Respirator Kit A1P2 R is a ready to use respiratory kit that includes 3M™ Half Mask 6200 large size and a filter set for replacement.
  • Ready to use respiratory kit with 3M™ Reusable Half Face Mask 6200L and 3M™ Filters offering A1P2 R protection
  • Supplied with 3M™ Filters offering A1P2 R protection
  • Bayonet connection fitting system to easily connect filters to mask
TRUSTED_SUSTAINABILITY
Specifications
Product Type Half Mask Respirator Kit
Size Large
Includes Half Mask 6200, filter set