missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Reusable Half Face Mask and P3 Particulate Filter Kit
This handy, ready-to-use kit includes a 3M™ Reusable Half Face Mask, a P3 filter and a pre-filter, all in a reusable storage box.
£770.00
Specifications
Product Type | Ready-to-Use Respirator Pack |
---|---|
Includes | 3M™ Reusable Half Face Mask, P3 Filter and Pre-Filter |
Quantity | 10/Cs. |
Product Code | Brand | Model | Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Size | Price | Quantity & Availability | ||||
17786983
|
3M™
7100265070 |
HF-3011 | Small |
£770.00
Pack of 10 |
Estimated Shipment: 02-07-2024
Log in to see stock. |
||||
17756993
|
3M™
7100265235 |
HF-3021 | Medium |
£770.00
Pack of 10 |
Estimated Shipment: 02-07-2024
Log in to see stock. |
||||
17796983
|
3M™
7100265071 |
HF-3031 | Large |
£770.00
Pack of 10 |
Estimated Shipment: 02-07-2024
Log in to see stock. |
||||
Description
- Includes 1 x 3M™ Reusable Half Face Mask, 1 x 3M™ Particulate Filter PF10 P3 R D DT-1135E, 1 x 3M™ Prefilter SC-6600 and 1 x 3M™ Prefilter Holder SC-6602
- Offers P3 particulate protection
- Comes in a useful, reusable storage box
Specifications
Ready-to-Use Respirator Pack | |
3M™ Reusable Half Face Mask, P3 Filter and Pre-Filter | |
10/Cs. |