missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RETSAT Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA552491
This item is not returnable.
View return policy
Description
Immunogen sequence: GVSVKKGHEL VNIYCPIVVS NAGLFNTYEH LLPGNARCLP GVKQQLGTVR PGLGMTSVFI CLRGTKEDLH LPSTNYYVYY DTDMDQAMER YVSMPREEAA EHIPLLFFAF PSAKDPTWE Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 76%.
Retinol saturase carrying out the saturation of the 13-14 double bond of all-trans-retinol to produce all-trans-13,14-dihydroretinol. Has activity toward all-trans-retinol as substrate. Does not use all-trans-retinoic acid nor 9-cis, 11-cis or 13-cis-retinol isomers as substrates. May play a role in the metabolism of vitamin A.
Specifications
| RETSAT | |
| Polyclonal | |
| Unconjugated | |
| RETSAT | |
| All-trans-13,14-dihydroretinol saturase; all-trans-retinol 13,14-reductase; PPAR-alpha-regulated and starvation-induced gene protein; PPSIG; retinol saturase; retinol saturase (all-trans-retinol 13,14-reductase); RETSAT; UNQ439/PRO872 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 54884 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6NUM9 | |
| RETSAT | |
| Recombinant protein corresponding to Human RETSAT. Recombinant protein control fragment (Product #RP-89961). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction