Learn More
Brady™ Retro Reflective All-weather Labels
Increase safety and visibility with the B-584 highly-reflective label material. Print labels almost anywhere high visibility is needed with a compact Brady™ handheld label printer.
£34.51 - £57.42
Specifications
Autoclavable Not Autoclavable
Certifications/Compliance 2011/65/EU, 2015/863
Coating None
Printable Sides Top
Print Technology Thermal Transfer
View More Specs
Products 8
Product Code Brand Dimensions (L x W) Color For Use With (Equipment) Price Quantity & Availability  
Product Code Brand Dimensions (L x W) Color For Use With (Equipment) Price Quantity & Availability  
17981940
View Documents
Brady™
170809
12.70 mm (W) x 6.10 m (L) White BMP41, BMP51, BMP53, M511, M410, M510
ONLINE EXCLUSIVE PRICE
£45.50
£34.51 / Pack of 1
Save £10.99
24% Off
View alternative products
Save Up to 
»
Estimated Shipment: 24-12-2026
Add to basket
17971950
View Documents
Brady™
170810
12.70 mm (W) x 6.10 m (L) Yellow BMP41, BMP51, BMP53, M511, M410, M510
ONLINE EXCLUSIVE PRICE
£40.70
£34.51 / Pack of 1
Save £6.19
15% Off
View alternative products
Save Up to 
»
Estimated Shipment: 08-01-2027
Add to basket
17971930
View Documents
Brady™
170811
19.05 mm (W) x 6.10 m (L) White BMP41, BMP51, BMP53, M511, M410, M510
ONLINE EXCLUSIVE PRICE
£42.25
£40.27 / Pack of 1
Save £1.98
4% Off
View alternative products
Save Up to 
»
Estimated Shipment: 24-12-2026
Add to basket
17931960
View Documents
Brady™
170812
19.05 mm (W) x 6.10 m (L) Yellow BMP41, BMP51, BMP53, M511, M410, M510
ONLINE EXCLUSIVE PRICE
£42.25
£40.27 / Pack of 1
Save £1.98
4% Off
View alternative products
Save Up to 
»
Estimated Shipment: 24-12-2026
Add to basket
17801769
View Documents
Brady™
170935
25.40 mm (W) x 6.10 m (L) White BMP51, BMP53, M511, M510
ONLINE EXCLUSIVE PRICE
£48.15
£45.91 / Pack of 1
Save £2.24
4% Off
View alternative products
Save Up to 
»
Estimated Shipment: 24-12-2026
Add to basket
17831759
View Documents
Brady™
170936
25.40 mm (W) x 6.10 m (L) Yellow BMP51, BMP53, M511, M510
ONLINE EXCLUSIVE PRICE
£48.15
£45.91 / Pack of 1
Save £2.24
4% Off
View alternative products
Save Up to 
»
Estimated Shipment: 24-12-2026
Add to basket
17861739
View Documents
Brady™
170937
38.10 mm (W) x 6.10 m (L) White BMP51, BMP53, M511, M510
ONLINE EXCLUSIVE PRICE
£60.15
£57.42 / Pack of 1
Save £2.73
4% Off
View alternative products
Save Up to 
»
Estimated Shipment: 24-12-2026
Add to basket
17851739
View Documents
Brady™
170938
38.10 mm (W) x 6.10 m (L) Yellow BMP51, BMP53, M511, M510
ONLINE EXCLUSIVE PRICE
£60.15
£57.42 / Pack of 1
Save £2.73
4% Off
View alternative products
Save Up to 
»
Estimated Shipment: 24-12-2026
Add to basket
Description
Description
  • Ideal for safety applications such as on personnel, fire suppression equipment, and first aid cabinets or anywhere extra visibility is required
  • Resistant to abrasions, weather, solvents and extreme temperatures
  • Pressure-sensitive acrylic adhesive withstands prolonged exposure to tough conditions
  • Able to withstand outdoor conditions for 4 to 6 years on average
Specifications
Specifications
Not Autoclavable
None
Thermal Transfer
Component and equipment labelling;Facility identification;General identification;Safety identification
1 Label(s) / Box
-40°F to 158°F
0.0055 in
20 ft.
0.5 in.
2011/65/EU, 2015/863
Top
Label
-40°C
1
-40°C to 70°C
0.14 mm
6.10 m