missing translation for 'onlineSavingsMsg'
Learn More

Retro-2 cycl, MedChemExpress

Product Code. 16425636
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16425636

Brand: Medchem Express HY114698

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Retro-2 cycl (RN 1-001) is a dihydroquinazolinone (DHQZ) inhibitor of retrograde trafficking. Retro-2 cycl (RN 1-001) inhibits JCPyV and HPV16 pseudovirus with IC50s of 54 μM and 160 μM, respectively. Antiviral agent.

Retro-2 cycl (RN 1-001) is a dihydroquinazolinone (DHQZ) inhibitor of retrograde trafficking. Retro-2 cycl (RN 1-001) inhibits JCPyV and HPV16 pseudovirus with IC50s of 54 μM and 160 μM, respectively. Antiviral agent.

Specifications

Chemical Name or Material Retro-2 cycl
Physical Form Solid
CAS 1429192-00-6
Color Off-White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C19H16N2OS
For Use With (Application) COVID-19-anti-virus
Synonym RN 1-001
Solubility Information DMSO : ≥ 125 mg/mL (390.13 mM)
SMILES O=C1C2=CC=CC=C2NC(N1C3=CC=CC=C3)C4=CC=C(S4)C
Molecular Weight (g/mol) 320.41
Formula Weight 320.41
Percent Purity 99.86%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.