missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Retnlb, Rat anti-Mouse, Clone: MRB 46L, Abnova™
Rat Monoclonal Antibody
Brand: Abnova MAB15884.100ug
This item is not returnable.
View return policy
Specifications
| Retnlb | |
| Monoclonal | |
| Unconjugated | |
| In PBS, pH 7.4. | |
| 9030012B21Rik/Fizz2/RELMbeta/Relmb/Xcp3 | |
| Rat | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2a κ |
| ELISA, Western Blot | |
| MRB 46L | |
| ELISA (1:2000-1:10000) Western Blot (1:2000-1:10000) The optimal working dilution should be determined by the end user. | |
| resistin like beta | |
| Retnlb | |
| Recombinant protein corresponding to mouse Retnlb. | |
| RUO | |
| 57263 | |
| For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction