missing translation for 'onlineSavingsMsg'
Learn More

Retinyl palmitate, MedChemExpress

Product Code. 16458746
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16458746 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16458746 Supplier Medchem Express Supplier No. HYB1384
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Retinyl palmitate is an ester of Retinol and is the major form of vitamin A found in the epidermis. Retinyl palmitate has been widely used in pharmaceutical and cosmetic formulations.

Retinyl palmitate is an ester of Retinol and is the major form of vitamin A found in the epidermis. Retinyl palmitate has been widely used in pharmaceutical and cosmetic formulations.

Specifications

Chemical Name or Material Retinyl palmitate
CAS 79-81-2
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C36H60O2
Quantity 100 mg
Synonym Vitamin A palmitate Retinol palmitate
Solubility Information Ethanol : 10 mg/mL (19.05 mM; Need ultrasonic)
SMILES CC(/C=C/C=C(/C=C/C1=C(CCCC(C)1C)C)C)=C\COC(CCCCCCCCCCCCCCC)=O
Molecular Weight (g/mol) 524.86
Formula Weight 524.86
Percent Purity 97.0%
Grade Research
Physical Form Liquid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.