missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Retinol Binding Protein RBP Antibody (SPM442), Novus Biologicals™
Mouse Monoclonal Antibody
£221.00 - £382.00
Specifications
| Antigen | Retinol Binding Protein RBP |
|---|---|
| Clone | SPM442 |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot 1-2 ug/mL, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1-2 ug/mL, Immunohistochemistry-Paraffin 1-2 ug/mL |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18458941
|
Novus Biologicals
NBP2-34362-0.02mg |
0.02 mg |
£221.00
0.02mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18478961
|
Novus Biologicals
NBP2-34362-0.1mg |
0.1 mg |
£382.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Retinol Binding Protein RBP Monoclonal antibody specifically detects Retinol Binding Protein RBP in Human, Mouse, Rat, Goat, Monkey, Rabbit samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| Retinol Binding Protein RBP | |
| 0.2 mg/mL | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cancer, Diabetes Research, Lipid and Metabolism | |
| 10 mM PBS with 0.05% BSA | |
| C, Cellular retinol-binding protein, CRABP-I, CRBP1CRBP-I, CRBPCellular retinol-binding protein I, CRBPI, RBPC, retinol binding protein 1, cellular, retinol-binding protein 1, retinol-binding protein 1, cellular | |
| Retinol binding protein purified from human plasma | |
| Primary | |
| Store at 4C. |
| SPM442 | |
| Western Blot 1-2 ug/mL, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1-2 ug/mL, Immunohistochemistry-Paraffin 1-2 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat, Goat, Monkey, Rabbit | |
| P02753, P09455, P50120, P02753 | |
| 5947 | |
| IgG1 κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title