missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Retinol Binding Protein RBP Antibody (OTI2H3), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73852PCP
This item is not returnable.
View return policy
Description
Retinol Binding Protein RBP Monoclonal specifically detects Retinol Binding Protein RBP in Human, Mouse, Rat samples. It is validated for Flow Cytometry.
Specifications
| Retinol Binding Protein RBP | |
| Monoclonal | |
| PerCP | |
| PBS with 0.05% Sodium Azide | |
| RBP1 | |
| Full length recombinant protein of human Retinol Binding Protein RBP (NP_002890) produced in HEK293T cells. | |
| 0.1 mL | |
| 5947.0 | |
| Store at 4°C in the dark. |
| Flow Cytometry | |
| OTI2H3 | |
| Flow Cytometry | |
| C, Cellular retinol-binding protein, CRABP-I, CRBP1CRBP-I, CRBPCellular retinol-binding protein I, CRBPI, RBPC, retinol binding protein 1, cellular, retinol-binding protein 1, retinol-binding protein 1, cellular | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction