missing translation for 'onlineSavingsMsg'
Learn More

Retinoic acid, 97%

Product Code. 11475290
Change view
Click to view available options
Quantity:
0.1 g
0.5 g
2 g
Unit Size:
0.1g
0.5g
2g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11475290 2 g 2g
11485290 0.1 g 0.1g
11465290 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11475290 Supplier Thermo Scientific Alfa Aesar Supplier No. 044540.04
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

All-trans-retinol, Vitamin A, retinol is a fat-soluble vitamin that binds retinoid receptors within cells. | CAS: 302-79-4 | C20H28O2 | 300.44 g/mol

Retinoic acid is used in dermatology drugs. It is used in the treatment of psoriasis, ichthyosis, follicular keratosis, acne, lichen planus, verrucous epidermal nevus and lichen psoriasis. It is also employed in promoting the proliferation and differentiation of epithelial cells. In addition to this, it exhibits some preventive effect on skin cancer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description

• All-trans-retinol, commonly known as Vitamin A, is a fat-soluble essential nutrient

• All-trans-retinol is involved in hormonal signaling via the retinoid receptors

• It is an important regulator of cell division and apoptosis

• Deficiency can lead to vision, bone, immune, and skin disorders

Applications

• Suitable for research on cell differentiation, embryonic development, immunology and more

• Laboratory grade quality, for research use only

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 302-79-4
Molecular Formula C20H28O2
Molecular Weight (g/mol) 300.44
MDL Number MFCD00001551
InChI Key SHGAZHPCJJPHSC-YCNIQYBTSA-N
Synonym retinoic acid, tretinoin, vitamin a acid, all-trans-retinoic acid, trans-retinoic acid, renova, airol, retin-a, vesanoid, atra
PubChem CID 444795
ChEBI CHEBI:15367
SMILES C\C(\C=C\C1=C(C)CCCC1(C)C)=C/C=C/C(/C)=C/C(O)=O

Specifications

Melting Point 180°C to 182°C
Quantity 2 g
Beilstein 2057223
Sensitivity Light sensitive
Merck Index 14,8165
Solubility Information Soluble in ethanol,acetate,dimethyl formamide,dimethyl sulfoxide,chloroform,methanol,acetone and dichloromethane. Insoluble in water and ether.
IUPAC Name (2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohexen-1-yl)nona-2,4,6,8-tetraenoic acid
Formula Weight 300.44
Percent Purity 98%
Physical Form Crystalline
Chemical Name or Material Retinoic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.