missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Retinoblastoma binding protein 1 Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-26616R
This item is not returnable.
View return policy
Description
Retinoblastoma binding protein 1 Polyclonal antibody specifically detects Retinoblastoma binding protein 1 in Human samples. It is validated for Western Blot
Specifications
| Retinoblastoma binding protein 1 | |
| Polyclonal | |
| Western Blot | |
| AT rich interactive domain 4A (RBP1-like), RBBP-1, RBP-1, RBP1ARID domain-containing protein 4A, retinoblastoma binding protein 1, Retinoblastoma-binding protein 1RBBP1AT-rich interactive domain-containing protein 4A | |
| The immunogen recognized by this antibody maps to a region between residue 900 and 950 of human AT-rich interactive domain-containing protein 4A using the numbering given in entry NP_002883.3 (GeneID 5926). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| DyLight 550 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 5926 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction