missing translation for 'onlineSavingsMsg'
Learn More

Retinoblastoma binding protein 1 Antibody, Alexa Fluor™ 750, Novus Biologicals™

Product Code. 18406416 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18406416 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18406416 Supplier Novus Biologicals Supplier No. NBP126616AF750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Retinoblastoma binding protein 1 Polyclonal antibody specifically detects Retinoblastoma binding protein 1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Retinoblastoma binding protein 1
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 750
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias AT rich interactive domain 4A (RBP1-like), RBBP-1, RBP-1, RBP1ARID domain-containing protein 4A, retinoblastoma binding protein 1, Retinoblastoma-binding protein 1RBBP1AT-rich interactive domain-containing protein 4A
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 900 and 950 of human AT-rich interactive domain-containing protein 4A using the numbering given in entry NP_002883.3 (GeneID 5926).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5926
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.