missing translation for 'onlineSavingsMsg'
Learn More

Retinoblastoma binding protein 1 Antibody (2G8), Novus Biologicals™

Product Code. 18393609 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18393609 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18393609 Supplier Novus Biologicals Supplier No. H00005926M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Retinoblastoma binding protein 1 Monoclonal antibody specifically detects Retinoblastoma binding protein 1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Retinoblastoma binding protein 1
Applications Western Blot, ELISA, Immunocytochemistry
Classification Monoclonal
Clone 2G8
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_002883
Gene Alias AT rich interactive domain 4A (RBP1-like), RBBP-1, RBP-1, RBP1ARID domain-containing protein 4A, retinoblastoma binding protein 1, Retinoblastoma-binding protein 1RBBP1AT-rich interactive domain-containing protein 4A
Host Species Mouse
Immunogen ARID4A (NP_002883, 1033 a.a. ~ 1139 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. AEESQEGLCERESANGFETNVASGTCSIIVQERESREKGQKRPSDGNSGLMAKKQKRTPKRTSAAAKNEKNGTGQSSDSEDLPVLDNSSKCTPVKHLNVSKPQKLAR
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5926
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.