missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Retigabine
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Retigabine is a KV7 (KCNQ) channel activator (EC50 values are 0.6 - 100 μM for KV7.1 - KV7.5). Anticonvulsant. Orally bioavailable.
Specifications
Specifications
| Chemical Name or Material | Ethyl [2-amino-4-[[(4-fluorophenyl)methyl]amino]phenyl]carbamate |
| CAS | 150812-12-7 |
| Quantity | 10 mg |
| Target | Voltage-gated Potassium Channel Activators |
| Molecular Formula | C16H18FN3O2 |
| Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction