missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Biosciences™ Reticulocyte Whole Blood Control (CE/IVD)
Click to view available options
Quantity:
200 Tests
Unit Size:
1 set
Description
- Tri-level, assayed hematology control designed to document and monitor values obtained from manual and automated reticulocyte counting methods.
- Contents: human erythrocytes and mammalian erythrocytes suspended in a plasma-like fluid with preservatives, in 3 levels: normal, moderately high, and very high.
Specifications
Specifications
| Content And Storage | Store vials upright, tightly capped, at 2°C to 8°C when not in use. Protect vials from overheating or freezing. |
| For Use With (Application) | Reticulocyte Count |
| For Use With (Equipment) | Flow Cytometer |
| Quantity | 200 Tests |
| Stability | Unopened vials are stable until the expiration date indicated on each vial and Assay Values sheet. Opened vials are stable for 21 days when handled properly. If sample preparation is a separate step before counting, count the prepared sample within 60 minutes after the minimum incubation time. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction