Learn More
Leica Microsystems Reticle, DM750
Click to view available options
For Use With (Equipment):
DM500/750 Microscope Eyepiece
Unit Size:
Each
Description
- Can be used for linear measurement, particle counting, point of reference etc.
- Suitable for eyepieces of DM500/750 microscopes.
Order Info
ForUseWithEquipment
Specifications
Specifications
| Type | Reticle |
| Diameter (Metric) | 10 mm |
| For Use With (Equipment) | DM500/750 Microscope Eyepiece |
| Quantity | 1 Ea. |