missing translation for 'onlineSavingsMsg'
Learn More

Retene, 97%

Product Code. 15505906 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15505906 250 mg 250mg
15595896 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15505906 Supplier Thermo Scientific Acros Supplier No. 455652500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 483-65-8 | C18H18 | 234.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 483-65-8
Molecular Formula C18H18
Molecular Weight (g/mol) 234.34
MDL Number MFCD00016358
InChI Key NXLOLUFNDSBYTP-UHFFFAOYSA-N
Synonym retene, reten, 1-methyl-7-isopropylphenanthrene, 7-isopropyl-1-methylphenanthrene, phenanthrene, 1-methyl-7-1-methylethyl, 1-methyl-7-1-methylethyl phenanthrene, unii-0w2d2e1p9q, phenanthrene, 7-isopropyl-1-methyl, ccris 3180, 7-isopropyl-1-methylphenanthren
PubChem CID 10222
IUPAC Name 1-methyl-7-propan-2-ylphenanthrene
SMILES CC(C)C1=CC=C2C(C=CC3=C(C)C=CC=C23)=C1

Specifications

Melting Point 97.0°C to 99.0°C
Boiling Point 108.0°C (10.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 250 mg
Solubility Information Solubility in water: negligible. Other solubilities: soluble in hot alochol, benzene, hot ether, carbon, disulfide
Formula Weight 234.34
Percent Purity 97%
Chemical Name or Material Retene
compliance-icons
Product Identifier
  • Retene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.