missing translation for 'onlineSavingsMsg'
Learn More

Scientific Industries Retaining Bands Kit for Rotator Genie

Product Code. 15883561
Click to view available options
For Use With (Equipment):
Rotator Genie
Unit Size:
Pack of 8
This item is not returnable. View return policy

Product Code. 15883561

Brand: Scientific Industries 0K2100901

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

This Retaining Bands Kit for Rotator Genie is for use with the Rotator Genie Shaker Platform.

  • Replacement elastic retaining bands (8) for Rotator Genie standard platform

Specifications

Product Type Retaining Bands
Weight (English) 1 lb.
Weight (Metric) 0.45 kg
Color Black
For Use With (Equipment) Rotator Genie
Packaging Quantity 8 Elastic Bands
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.