missing translation for 'onlineSavingsMsg'
Learn More

Hirschmann™ Retainer Ring

Product Code. 10044270
Change view
Click to view available options
Color:
Chrome
Gray
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
10044270 Chrome Each
10431783 Gray Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10044270 Supplier Hirschmann™ Supplier No. 9903802
Add to basket
Add to basket
This item is not returnable. View return policy

Spare part retainer ring chromated for pipetus™ pipetholder.

  • Spare part retainer ring chromated for pipetus™ pipetholder.

Specifications

Color Chrome
Type Ring Union Nut
For Use With (Equipment) Pipetus, Pipetus Junior, and Pipetus Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.