missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Retainer & Base Set (Standard) for 3500/3500xL Genetic Analyzers, 96 well

Product Code. 15853560 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
4 Sets
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15853560 4 Sets Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15853560 Supplier Applied Biosystems™ Supplier No. 4410228
Discount Lab Supplies
Use promo code 28170
View all promotions
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This 96-well plate Retainer and Base Set is for use with Fast (0.1mL) 96-well samples pates on the 3500 series Genetic Analyzers.

This 96-well plate Retainer and Base Set is for use with standard 96-well sample plates on the 3500 series Genetic Analyzers.

For Research Use Only. Not for use in diagnostics procedures.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage This product includes 4 sets of retainers and bases.
Description Retainer & Base Set (Standard), 96-well Plate, For 3500/3500XL Genetic Analyzers, Room Temperature
Format 96-well Plate
For Use With (Equipment) 3500 Genetic Analyzer, 3500xL Genetic Analyzer
For Use With (Application) Sequencing
Product Type Base Set
Quantity 4 Sets
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.