missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ 3500 Dx Series 96-Well Retainer & Base Set

Product Code. 17286483 Shop All Applied Biosystems Products
Click to view available options
Quantity:
4 Sets
Unit Size:
Pack of 4
This item is not returnable. View return policy

Product Code. 17286483

Brand: Applied Biosystems™ 4410227

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This is a 96-well retainer & base set (standard) designed for use with the 3500 Dx and 3500xL Dx genetic analyzers.

This is a retainer and base set (standard) designed for septa retention in 8-tube strip and 96-well plate formats for use with the 3500 Dx and 3500xL Dx genetic analyzers.

Note: The 3500 Dx and 3500xL Dx genetic analyzers and consumables are for distribution and use in select countries only.

  • This is a 96-well retainer & base set (standard) designed for use with the 3500 Dx⁄3500xL Dx Genetic Analyzer.
  • Septa and plate base⁄retainer kits for 8 tube strip, 96 and 384-well plates used with the 3500 Dx Series Genetic Analyzers.
  • The 3500 Dx Series Systems and consumables are for distribution and use in select countries only.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Ambient storage.
Each package contains 4 sets.
For Use With (Equipment) 3500xL Dx Genetic Analyzer, 3500 Dx Genetic Analyzer, 3500xL Genetic Analyzer, 3500 Genetic Analyzer
For Use With (Application) Sequencing
Plate Compatibility 96-Well Microtiter Plates, MicroAmp™ Optical 96-Well Reaction Plates
Product Type Base Set
Quantity 4 Sets
Shipping Condition Room Temperature

For In Vitro Diagnostic Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.