missing translation for 'onlineSavingsMsg'
Learn More

Eurofins DiscoverX Ret Protein, Active

Product Code. 17320364
Encompass_Preferred
Click to view available options
Quantity:
10 μg
250 μg
Unit Size:
10µg
250µg
This item is not returnable. View return policy

Product Code. 17320364

Brand: Eurofins DiscoverX 14570

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Active, N-Terminal GST-tagged, recombinant, human Ret amino acids 658-end, for use in Kinase Assays.

The RET (rearranged during transfection) receptor tyrosine kinase is a classic example of phenotype heterogeneity. Gain-of-function mutations of RET are associated with human cancer and multiple endocrine neoplasia type 2 (MEN 2A, MEN 2B, etc.) Loss-of-function mutations of RET cause Hirschspung disease or colonic aganglionosis).

GenBank NM_000323

Specifications

Formulation mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 150 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution.
Molecular Weight (g/mol) 79.2 kDa
Quantity 10 μg
Species Human
Recombinant Recombinant
Protein Tag N-terminal GST
Expression System Recombinant enzyme expressed in Sf21 insect cells
Protein Form Truncated
Purity or Quality Grade ≥58%
Protein Ret, active
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.