missing translation for 'onlineSavingsMsg'
Learn More

Ret, Mouse anti-Human, Clone: RET/2663, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-79761-20UG

 View more versions of this product

Product Code. 18298097

  • £203.00 / 20µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Ret Monoclonal specifically detects Ret in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Ret
Monoclonal
0.2 mg/mL
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array
P07949, P07949
RET
Recombinant fragment (around aa 702-848) of human Ret protein (exact sequence is proprietary)
20 μg
Cancer, Oncogenes, Phospho Specific, Protein Kinase, Protein Phosphatase, Signal Transduction, Tumor Suppressors
5979
Store at 4°C.
IgG2b κ
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array
RET/2663
Unconjugated
10 mM PBS with 0.05% BSA
CDHF12MTC1, CDHR16, EC 2.7.10, Hirschsprung disease 1, Hirschsprung disease), MEN2A, MEN2B, multiple endocrine neoplasia and medullary thyroid carcinoma 1, ret proto-oncogene, RET transforming sequence
Mouse
Protein A or G purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.