Learn More
RET (extracellular) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA5111870
Description
Reconstitute antibody with 50 μL of double distilled water (DDW). The reconstituted solution can be stored at 4 degrees C for up to 1 week. For longer periods, small aliquots should be stored at -20 degrees C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min). Peptide sequence is: CEWRQGDGKG ITR Homology: Mouse amino acid residues identical; rat - 12/13 amino acid residues identical.
RET is a member of the cadherin superfamily, encodes one of the receptor tyrosine kinases which are cell-surface molecules that transduce signals for cell growth and differentiation. RET plays a crucial role in neural crest development, can undergo oncogenic activation in vivo and in vitro by cytogenetic rearrangement. Mutations in the RET gene are associated with the disorders multiple endocrine neoplasia, type IIA, multiple endocrine neoplasia, type IIB, Hirschsprung disease, and medullary thyroid carcinoma. Two transcript variants encoding different isoforms have been found for the RET gene. Additional transcript variants have been described for RET but their biological validity has not been confirmed.
Specifications
| RET (extracellular) | |
| Polyclonal | |
| Unconjugated | |
| RET | |
| Cadherin family member 12; CDHF12; CDHR16; CG1061; CG14396; c-Ret; Dmel\CG14396; Dmel_CG14396; DmHD-59; DmRet; dret; D-ret; EC 2.7.10.1; Extracellular cell-membrane anchored RET cadherin 120 kDa fragment; HD-59; HSCR1; kinase Ret; MEN2; MEN2A; MEN2B; MTC1; OTTHUMP00000216967; Proto-oncogene c-Ret; Proto-oncogene tyrosine-protein kinase receptor Ret; PTC; RET; RET51; RET9; RET-ELE1; Reto; Soluble RET kinase fragment | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 19713, 24716, 5979 | |
| -20°C | |
| Lyophilized |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunomicroscopy, Western Blot | |
| 0.8 mg/mL | |
| PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
| G3V9H8, P07949, P35546 | |
| RET | |
| Peptide corresponding to amino acid residues 541-553 of human Ret (Extracellular, N-terminus) | |
| 50 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.