missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Ret Antibody (Regeneron patent anti-RET), Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28578-1mg
This item is not returnable.
View return policy
Description
Ret Monoclonal antibody specifically detects Ret in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| Ret | |
| Monoclonal | |
| Unconjugated | |
| P07949 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| Regeneron patent anti-RET | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| CDHF12MTC1, CDHR16, EC 2.7.10, Hirschsprung disease 1, Hirschsprung disease), MEN2A, MEN2B, multiple endocrine neoplasia and medullary thyroid carcinoma 1, ret proto-oncogene, RET transforming sequence | |
| c-RET | |
| 1 mg | |
| Cancer, Oncogenes, Phospho Specific, Protein Kinase, Protein Phosphatase, Signal Transduction, Tumor Suppressors | |
| 5979 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction