missing translation for 'onlineSavingsMsg'
Learn More

Resveratrol, MedChemExpress

Product Code. 16481806
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 mg
500 mg
Unit Size:
200mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16481806 500 mg 500mg
16411226 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16481806 Supplier Medchem Express Supplier No. HY16561

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Resveratrol (trans-Resveratrol; SRT501), a natural polyphenolic phytoalexin that possesses anti-oxidant, anti-inflammatory, cardioprotective, and anti-cancer properties.

Resveratrol (SRT 501) has a wide spectrum of targets including mTOR, JAK, β-amyloid, Adenylyl cyclase, IKKβ, DNA polymerase. Resveratrol also is a specific SIRT1 activator. Resveratrol is a potent pregnane X receptor (PXR) inhibitor. Resveratrol is an Nrf2 activator, ameliorates aging-related progressive renal injury in mice model. Resveratrol increases production of NO in endothelial cells.

Specifications

Chemical Name or Material Resveratrol
CAS 501-36-0
Color Off-White
Purity Grade Notes Research
Molecular Formula C14H12O3
Quantity 500 mg
Synonym trans-Resveratrol SRT501
Solubility Information DMSO : 100 mg/mL (438.14 mM; Need ultrasonic) ∣Ethanol : 50 mg/mL (219.07 mM; Need ultrasonic)
SMILES OC1=CC=C(/C=C/C2=CC(O)=CC(O)=C2)C=C1
Molecular Weight (g/mol) 228.24
Formula Weight 228.24
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.