missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Resuspension Solution for MagJET Plasmid DNA Kit

The Thermo Scientific MagJET Plasmid DNA Kit is designed for fast and efficient purification of plasmid DNA from overnight cultures of E. coli cultures. The kit utilizes magnetic bead technology, enabling high yields and robust performance.

Brand:  Thermo Scientific™ R2581

Code : 88

Additional Details : Weight : 0.07000kg

 View more versions of this product

Product Code. 13585220

  • £50.50 / 25mL
Estimated Shipment: 21-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Thermo Scientific MagJET Plasmid DNA Kit is designed for fast and efficient purification of plasmid DNA from overnight cultures of E. coli cultures. The kit utilizes magnetic bead technology, enabling high yields and robust performance. High binding capacity, uniform particle size, and rapid magnetic response of MagJET magnetic beads makes the technology ideal for high throughput automatic nucleic acid purification, as well as for manual purification by low sample throughput users. The resulting high-quality DNA is free of proteins, nucleases and other contaminants or inhibitors.

Specifications

Specifications

Suspension Buffer
MagJET
25 mL
Kit
Resuspension Solution
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.