missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Resuspension Solution for GeneJET Plasmid Maxiprep Kit

Product Code. 15564414 Shop All Thermo Scientific Products
Click to view available options
Quantity:
240 mL
Unit Size:
240mL
This item is not returnable. View return policy

Product Code. 15564414

Brand: Thermo Scientific™ R1581

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Thermo Scientific GeneJET Resuspension Buffer is a component of the GeneJET Plasmid Maxiprep Kit and may be purchased separately.

Thermo Scientific GeneJET Resuspension Buffer is a component of the GeneJET Plasmid Maxiprep Kit (Cat. No. K0491/K0492) and may be purchased separately.

Thermo Scientific GeneJET Resuspension Buffer is a component of the GeneJET Plasmid Maxiprep Kit (Cat. No. K0491/K0492) and may be purchased separately.
TRUSTED_SUSTAINABILITY

Specifications

Buffer Suspension Buffer
Format Kit
Product Line GeneJET
Product Type Resuspension Solution
Quantity 240 mL

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.