missing translation for 'onlineSavingsMsg'
Learn More

Restek™ Vacuum Vu-Union Connector for GC/MS Applications

Product Code. 13091770
Encompass_Preferred
Click to view available options
Packaging Quantity:
3/Pk.
This item is not returnable. View return policy

Product Code. 13091770

Brand: Restek™ 20428

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

For use with Vacuum Vu-Union Connector for GC-MS applications.

  • Fits column ODs from 0.33 to 0.74mm (Restek 0.1 to 0.53mm I.D.)
  • Quantity: 3/Pk.

Specifications

Type Replacement Insert
For Use With (Application) Use with Vacuum Vu-Union Connector for GC-MS Applications
Packaging Quantity 3/Pk.
For Use With Fits Column ODs From 0.33 to 0.74 mm (Restek 0.1 to 0.53 mm ID)
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.