Learn More
Restek™ Universal Press-Tight™ Connectors: Universal Connector
Product Code. 13081750
Encompass_Preferred
Click to view available options
Packaging Quantity:
25 Pack
5 Pack
Unit Size:
Pack of 5
Product Code. 13081750
Brand: Restek™ 20401
Only null left
Add to basket
Add to basket
Connect analytical columns, guard columns, transfer lines or detectors
  • Fit column outer diameters 0.33 to 0.74mm (Restek inside diameters 0.1 to 0.53mm)
  • Deactivated Press-Tight connectors for better recovery of polar and nonpolar compounds
  • Angle approximates the curvature of a capillary column, reducing the strain on column-end connections
Specifications
Type Connector
Packaging Quantity 25 Pack