Learn More
Graphite Ferrule for Uniliner Sleeve Adapter from Restek™
Click to view available options
Quantity:
5/Pk.
Unit Size:
Pack of 5
Description
- Reduces solvent tailing
- 0.53mm ID version can be used in the direct (DI) or on-column (OC) injection mode
- Incorporates a gentle taper that seals the column and reduces dead volume in direct injection mode
Specifications
Specifications
| Quantity | 5/Pk. |
| Product Type | Ferrules |
| For Use With (Equipment) | Uniliner Sleeve Adapters |