missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Uniliner Liners for 1/4-Inch Packed Injection Port Conversion from Restek™
For 0.25 in. packed injection port conversion.
Brand: Restek 20320
Code : Z3
Additional Details : Weight : 1.00000kg
Description
- Reduces solvent tailing
- 0.53mm ID version can be used in the direct (DI) or on-column (OC) injection mode
- Incorporates a gentle taper that seals the column and reduces dead volume in direct injection mode
Specifications
Cyclo-Uniliner; Direct Injection Only | |
5/Pk. | |
0.5 cm |
Cyclo UniLiner Liner | |
0.32, 0.53 mm | |
0.25 in. Packed Injection Port Conversion, 0.32 and 0.53 mm I.D. Column Direct Injection Only |