Learn More
Restek™ Tubing Reamer
Product Code. 13071570
Encompass_Preferred
Click to view available options
:
Tubing Reamer
Product Code. 13071570
Brand: Restek™ 20134
Only null left
Add to basket
Add to basket
Removes burrs from stainless steel tubing
  • For 6.4 or 3.2mm (1/4 or 1/8 in.) tubing
  • Nonslip safety design
Specifications
Type Tubing Reamer