Learn More
Restek™ Tubing Bender
Product Code. 10440605
Encompass_Preferred
Click to view available options
:
Tubing Bender
Product Code. 10440605
Brand: Restek™ 23009
Assists in making accurate left-hand, right-hand or offset bends
Triple-head tool bends 3.18, 4.76 and 6.35mm (1/8, 3/16 and 1/4 in.) tubing
Specifications
Type Tubing Bender