Learn More
Thermolite™ Septa from Restek™
Precision molding ensures consistent, accurate fit
£76.84
Product Code | Brand | Diameter (English) Outer | Diameter (Metric) Outer | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (English) Outer | Diameter (Metric) Outer | Quantity | Price | Quantity & Availability | |||
10480587
|
Restek
27142 |
0.43 in. | 11 mm | 50/Pk. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
Precision molding ensures consistent, accurate fit
- Usable to 340°C (644°F) inlet temperature
- Excellent puncturability
- Preconditioned and ready to use
- Packaged in ultra-clean blister packs