Learn More
Restek™ Syringe Adapter Kit
Product Code. 13106852
Encompass_Preferred
Change view
Click to view available options
Type:
Syringe Adapter Kit
Product Code. Type
13106852 Syringe Adapter Kit
1 options
Product Code. 13106852 Supplier Restek™ Supplier No. 21118
Estimated Shipment: 07-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Includes
One nickel-plated brass 1/4 in. NPT to female luer fitting, which can be used with an A-2 Luer syringe, One stainless steel, 1/4 in. NPT x 1/8 in. compression fitting with septum (can be used with any syringe needle).
For Single-Stage VOC Regulator
  • Use to withdraw sample from a high-pressure cylinder after pressure reduction through the high-purity VOC single-stage regulator
Specifications
Type Syringe Adapter Kit
For Use With (Application) Use to withdraw sample from a high pressure cylinder after pressure reduction through the high purity VOC single stage regulator
For Use With (Equipment) VOC single stage regulators
Includes One Nickel-plated Brass 0.25 in. NPT to female luer fitting, which can be used with an A-2 Luer syringe, one Stainless Steel, 0.25 in. NPT x 0.125 in. compression fitting with septum (can be used with any syringe needle)