Learn More
Restek™ Super-Clean Gas Trapping System for LC/MS
Product Code. p-3218366
Encompass_Preferred
Click to view available options
Type:
Gas Trapping System
Hydrocarbon Filter
Unit Size:
Each
Product Code. 13121452
Brand: Restek™ 22061
Quick-change cartridge system removing hydrocarbons efficiently from nitrogen
  • Changing filters is quick and easy
  • Up to 20L of hydrocarbon-free nitrogen per minute
  • Filters connected in parallel to handle high flows of LC/MS
Specifications
Type Hydrocarbon Filter
For Use With (Equipment) Super-Clean Gas Trapping System